• USA Home
  • 468398 - 1-Chloro-4-(trimethylsilyl)benzene

EMAIL THIS PAGE TO A FRIEND
468398 Sigma-Aldrich

1-Chloro-4-(trimethylsilyl)benzene

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.509 (lit.)
bp   98-100 °C/20 mmHg (lit.)
density   0.995 g/mL at 25 °C (lit.)
SMILES string   C[Si](C)(C)c1ccc(Cl)cc1
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?