• USA Home
  • 470686 - Lithium tetrakis(dimethylamino)aluminate

EMAIL THIS PAGE TO A FRIEND
470686 Sigma-Aldrich

Lithium tetrakis(dimethylamino)aluminate

97%

Purchase

Properties

assay   97%
mp   >255 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1759 8 / PGII
WGK Germany 
3
Flash Point(F) 
73.4 °F
Flash Point(C) 
23 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?