• USA Home
  • 47079 - Boc-(R)-α-benzyl-Pro-OH

EMAIL THIS PAGE TO A FRIEND
47079 Aldrich

Boc-(R)-α-benzyl-Pro-OH

≥99.0%

Synonym: (R)-2-Benzyl-1-Boc-2-pyrrolidinecarboxylic acid, Boc-α-benzyl L-proline

Purchase

Properties

assay   ≥99.0%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Proline Derivatives and Analogs

Proline is a non-polar proteinogenic amino acid that forms a tertiary amide when incorporated into peptides. It does not have a hydrogen on the amide group and therefore cannot act as a hydrogen bond...
Chemfiles Volume 5 Article 12
Keywords: Adhesion, Biochemistry, Catalysis, Deposition, Hydroxylations, Isomerizations, Nucleic acid hybridization, Pharmaceutical, Reductions, Substitutions, Type

Proline Derivatives

Proline is a non-polar, natural amino acid that forms a tertiary amide when incorporated into peptides. Thus, it is the only proteinogenic amino acid that does not act as a hydrogen bond donor in a p...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 8.
Keywords: Asymmetric synthesis, Chemfiles, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?