• USA Home
  • 471097 - Diethyl (α-aminobenzyl)phosphonate hydrochloride

EMAIL THIS PAGE TO A FRIEND
471097 Sigma-Aldrich

Diethyl (α-aminobenzyl)phosphonate hydrochloride

97%

Synonym: (Aminophenylmethyl)phosphonic acid diethyl ester hydrochloride, NSC 113536

  • CAS Number 16656-50-1

  • Linear Formula C6H5CH(NH2)P(O)(OC2H5)2 · HCl

  • Molecular Weight 279.70

  •  MDL number MFCD00101298

  •  PubChem Substance ID 24870697

  •  NACRES NA.22

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Horner-Wadsworth-Emmons Reagents, Olefination, Synthetic Reagents More...
Quality Level   100
assay   97%
reaction suitability   reaction type: C-C Bond Formation
mp   191 °C (dec.) (lit.)
SMILES string   Cl.CCOP(=O)(OCC)C(N)c1ccccc1
InChI   1S/C11H18NO3P.ClH/c1-3-14-16(13,15-4-2)11(12)10-8-6-5-7-9-10;/h5-9,11H,3-4,12H2,1-2H3;1H
InChI key   XMRSFMWMNXQRAP-UHFFFAOYSA-N

Description

Application

Reactant for:
• Synthesis of phosphonopeptides
• reaction with phosphorous pentachloride

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?