• USA Home
  • 472816 - 4-O-Acetyl-3,6-di-O-(tert-butyldiphenylsilyl)-D-glucal

EMAIL THIS PAGE TO A FRIEND
472816 Sigma-Aldrich

4-O-Acetyl-3,6-di-O-(tert-butyldiphenylsilyl)-D-glucal

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C13-C60, Carbohydrate Chemistry, Carbohydrate Derivatives,
optical activity   [α]25/D +7°, c = 1.5 in chloroform
impurities   3-5% ethyl acetate to induce mobility
SMILES string   CC(=O)O[C@@H]1[C@@H](CO[Si](c2ccccc2)(c3ccccc3)C(C)(C)C)OC=C[C@H]1O[Si](c4ccccc4)(c5ccccc5)C(C)(C)C
InChI   1S/C40H50O7Si2/c1-30(42)46-37(36(44)29-45-48(39(2,3)4,31-20-12-8-13-21-31)32-22-14-9-15-23-32)38(35(43)28-41)47-49(40(5,6)7,33-24-16-10-17-25-33)34-26-18-11-19-27-34/h8-28,35-38,43-44H,29H2,1-7H3/t35-,36+,37+,38+/m0/s1
InChI key   TVXPYOJFXNMPNJ-ZPAWJIGKSA-N

Description

Packaging

100, 500 mg in glass bottle

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?