• USA Home
  • 472875 - Tetra-O-acetyl-6,6′-di-O-(tert-butyldiphenylsilyl)-D-lactal

EMAIL THIS PAGE TO A FRIEND
472875 Sigma-Aldrich

Tetra-O-acetyl-6,6′-di-O-(tert-butyldiphenylsilyl)-D-lactal

97%

Purchase

Properties

assay   97%
optical activity   [α]20/D −20°, c = 1 in chloroform
mp   67-70 °C (lit.)
SMILES string   CC(=O)O[C@@H]1C=CO[C@H](CO[Si](c2ccccc2)(c3ccccc3)C(C)(C)C)[C@@H]1OC4O[C@H](CO[Si](c5ccccc5)(c6ccccc6)C(C)(C)C)[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]4OC(C)=O

Description

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?