• USA Home
  • 473057 - (R)-(+)-3-Hydroxy-5-oxo-1-cyclopentene-1-heptanoic acid

EMAIL THIS PAGE TO A FRIEND
473057 Sigma-Aldrich

(R)-(+)-3-Hydroxy-5-oxo-1-cyclopentene-1-heptanoic acid

96%

Purchase

Properties

assay   96%
optical activity   [α]22/D +17°, c = 1 in chloroform
mp   40-43 °C (lit.)
SMILES string   O[C@@H]1CC(=O)C(CCCCCCC(O)=O)=C1
InChI   1S/C12H18O4/c13-10-7-9(11(14)8-10)5-3-1-2-4-6-12(15)16/h7,10,13H,1-6,8H2,(H,15,16)/t10-/m0/s1
InChI key   IXOFUWJRSYPKSX-JTQLQIEISA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?