• USA Home
  • 473103 - Lithium-6Li chloride

EMAIL THIS PAGE TO A FRIEND
473103 Sigma-Aldrich

Lithium-6Li chloride

95 atom % 6Li

Purchase

Properties

InChI Key   KWGKDLIKAYFUFQ-HCMAANCNSA-M
isotopic purity   95 atom % 6Li
mp   605 °C (lit.)
mass shift   M-1

Description

Packaging

Packaged in screw-cap bottles

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?