• USA Home
  • 473855 - 1,6-Diiodoperfluorohexane

EMAIL THIS PAGE TO A FRIEND
473855 Sigma-Aldrich

1,6-Diiodoperfluorohexane

96%

Synonym: Dodecafluoro-1,6-diiodohexane, Perfluoro-1,6-diiodohexane

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
assay   96%
bp   173-174 °C (lit.)
mp   25-30 °C (lit.)
SMILES string   FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
InChI   1S/C6F12I2/c7-1(8,3(11,12)5(15,16)19)2(9,10)4(13,14)6(17,18)20
InChI key   JOQDDLBOAIKFQX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?