• USA Home
  • 474258 - 3,3,4,4,5,6,6,6-Octafluoro-5-(trifluoromethyl)hexyl methacrylate

EMAIL THIS PAGE TO A FRIEND
474258 Aldrich

3,3,4,4,5,6,6,6-Octafluoro-5-(trifluoromethyl)hexyl methacrylate

97%

  • CAS Number 65195-44-0

  • Linear Formula H2C=C(CH3)CO2CH2CH2CF2CF2CF(CF3)2

  • Molecular Weight 382.17

  •  MDL number MFCD00236096

  •  PubChem Substance ID 24870975

Purchase

Properties

assay   97%
contains   100 ppm 4-tert-butylcatechol as inhibitor
refractive index   n20/D 1.354 (lit.)
bp   199 °C(lit.)
density   1.481 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?