• USA Home
  • 474274 - 3,3,4,4,5,5,6,6,7,8,8,8-Dodecafluoro-7-(trifluoromethyl)octyl methacrylate

EMAIL THIS PAGE TO A FRIEND
474274 Sigma-Aldrich

3,3,4,4,5,5,6,6,7,8,8,8-Dodecafluoro-7-(trifluoromethyl)octyl methacrylate

97%

  • CAS Number 50836-66-3

  • Linear Formula H2C=C(CH3)CO2CH2CH2(CF2)4CF(CF3)2

  • Molecular Weight 482.19

  •  MDL number MFCD00236098

  •  PubChem Substance ID 24870979

Purchase

Properties

assay   97%
contains   100 ppm 4-tert-butylcatechol as inhibitor
refractive index   n20/D 1.347 (lit.)
bp   83-86 °C/0.85 mmHg (lit.)
density   1.567 g/mL at 25 °C (lit.)
SMILES string   CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(C(F)(F)F)C(F)(F)F
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?