• USA Home
  • 474371 - 3,3,4,4,5,5,6,6,7,8,8,8-Dodecafluoro-7-(trifluoromethyl)octyl acrylate

EMAIL THIS PAGE TO A FRIEND
474371 Sigma-Aldrich

3,3,4,4,5,5,6,6,7,8,8,8-Dodecafluoro-7-(trifluoromethyl)octyl acrylate

97%

  • CAS Number 50836-65-2

  • Linear Formula H2C=CHCO2CH2CH2(CF2)4CF(CF3)2

  • Molecular Weight 468.16

  •  MDL number MFCD00236111

  •  PubChem Substance ID 24870996

Purchase

Properties

assay   97%
contains   100 ppm 4-tert-butylcatechol as inhibitor
refractive index   n20/D 1.341 (lit.)
bp   216 °C (lit.)
density   1.607 g/mL at 25 °C (lit.)
SMILES string   FC(F)(F)C(F)(C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C
InChI   1S/C12H7F15O2/c1-2-5(28)29-4-3-6(13,14)8(16,17)10(20,21)9(18,19)7(15,11(22,23)24)12(25,26)27/h2H,1,3-4H2
InChI key   UZNPGWWLKHTVQQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?