• USA Home
  • 474495 - 4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluoro-2-hydroxynonyl acrylate

EMAIL THIS PAGE TO A FRIEND
474495 Aldrich

4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluoro-2-hydroxynonyl acrylate

96%

Purchase

Properties

assay   96%
contains   100 ppm 4-tert-butylcatechol as inhibitor
refractive index   n20/D 1.358(lit.)
bp   250 °C(lit.)
density   1.54 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?