• USA Home
  • 474657 - 3,5-Difluorosalicylaldehyde

EMAIL THIS PAGE TO A FRIEND
474657 Sigma-Aldrich

3,5-Difluorosalicylaldehyde

97%

Purchase

Description

Packaging

1 g in glass bottle

Application

3,5-Difluorosalicylaldehyde may be used in the synthesis of Ni[CpFe(η5-C5H4)-C(O)CH=C(CH3)N-o-C6H4N=CH-(2-O,3,5-F2-C6H2)] (Cp = cyclopentyl) and Zn(salphenazine) complex.

General description

3,5-Difluorosalicylaldehyde has been synthesized by employing 2,4-difluorophenol as a starting reagent.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?