EMAIL THIS PAGE TO A FRIEND
475106 Sigma-Aldrich

Equilin benzoate

97%

  • CAS Number 6030-80-4

  • Empirical Formula (Hill Notation) C25H24O3

  • Molecular Weight 372.46

  •  MDL number MFCD00468046

  •  PubChem Substance ID 24871079

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Complex Molecules
InChI Key   UIXFQXUDHNMOTI-KJWPAHLWSA-N
assay   97%
optical activity   [α]22/D +225°, c = 1 in chloroform
mp   193-196 °C (lit.)

Description

Packaging

50 mg in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KD7875000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?