• USA Home
  • 475483 - 6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one

EMAIL THIS PAGE TO A FRIEND
475483 Sigma-Aldrich

6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one

98%

  • CAS Number 26518-76-3

  • Empirical Formula (Hill Notation) C10H8ClNO3

  • Molecular Weight 225.63

  •  MDL number MFCD01321312

  •  PubChem Substance ID 24871135

Purchase

Properties

Related Categories Benzoxazines, Building Blocks, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks More...
InChI Key   MIAHXWVABDHISZ-UHFFFAOYSA-N
assay   98%
mp   228-230 °C (lit.)

Description

Packaging

25 g in glass bottle

Application

6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one may be used in the preparation of [2-(3-oxo-3,4-dihydro-2H-benzo[1,4]oxazin-6-carbonyl)-1H-indol-3-yl]acetic acids and N-alkyl 6-[2-(4,6-diphenylpyridyl)]-2H-[1,4]benzoxazin-3-one.

General description

6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one is a heterocyclic building block.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?