• USA Home
  • 475653 - Neopentyl glycol acrylate benzoate

EMAIL THIS PAGE TO A FRIEND
475653 Sigma-Aldrich

Neopentyl glycol acrylate benzoate

  • CAS Number 66671-22-5

  • Linear Formula H2C=CHCO2CH2C(CH3)2CH2O2CC6H5

  • Molecular Weight 262.30

  •  MDL number MFCD01321317

Purchase

Properties

contains   200 ppm monomethyl ether hydroquinone as inhibitor
refractive index   n20/D 1.502 (lit.)
density   1.08 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
230 °F

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?