• USA Home
  • 476110 - Bis(2-hydroxyphenyl)methane

EMAIL THIS PAGE TO A FRIEND
476110 Sigma-Aldrich

Bis(2-hydroxyphenyl)methane

95%

Synonym: 2,2′-Methylenediphenol

Purchase

Properties

InChI Key   MQCPOLNSJCWPGT-UHFFFAOYSA-N
assay   95%
mp   113-118 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B46808 Bis(2-hydroxyphenyl)methane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?