EMAIL THIS PAGE TO A FRIEND
47626 Sigma-Aldrich

Fmoc-Gln-OH

≥95.0% (T)

Synonym: Fmoc-L-glutamine

  • CAS Number 71989-20-3

  • Empirical Formula (Hill Notation) C20H20N2O5

  • Molecular Weight 368.38

  •  Beilstein/REAXYS Number 4722773

  •  EC Number 276-254-3

  •  MDL number MFCD00037137

  •  PubChem Substance ID 57651046

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Glutamine, Peptide Synthesis and Peptide Chemistry More...
InChI Key   IZKGGDFLLNVXNZ-KRWDZBQOSA-N
assay   ≥95.0% (T)
optical activity   [α]20/D −18.5±1.5°, c = 1% in DMF
mp   220 °C (dec.) (lit.)
storage temp.   2-8°C

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?