EMAIL THIS PAGE TO A FRIEND
476366 Sigma-Aldrich

o-Tolidine sulfone

Mixture of 2,8- and 4,8-dimethyl isomers, 99%

Synonym: 3,7-Diamino-2(4),8-dimethyldibenzothiophene 5,5-dioxide

  • CAS Number 55011-44-4

  • Empirical Formula (Hill Notation) C14H14N2O2S

  • Molecular Weight 274.34

  •  PubChem Substance ID 24871245

Purchase

Properties

assay   99%
mp   >350 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
UN 3077 9/PG 3
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?