• USA Home
  • 477591 - PSS-(3-Chloropropyl)-Heptacyclopentyl substituted

EMAIL THIS PAGE TO A FRIEND
477591 Sigma-Aldrich

PSS-(3-Chloropropyl)-Heptacyclopentyl substituted

Synonym: 1-(3-Chloropropyl)-3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, 3-Chloropropyl-POSS®

  • CAS Number 308103-62-0

  • Empirical Formula (Hill Notation) C38H69ClO12Si8

  • Molecular Weight 978.08

  •  MDL number MFCD01321336

  •  PubChem Substance ID 24871391

Purchase

Properties

mp   >350 °C (lit.)
SMILES string   ClCCC[Si]12O[Si]3(O[Si]4(O[Si](O1)(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)C6CCCC6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)C%11CCCC%11)C%12CCCC%12

Description

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?