• USA Home
  • 477605 - PSS-Glycidyl-Heptacyclopentyl substituted

EMAIL THIS PAGE TO A FRIEND
477605 Sigma-Aldrich

PSS-Glycidyl-Heptacyclopentyl substituted

95%

Synonym: 1,3,5,7,9,11,13-Heptacyclopentyl-15-glycidylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Glycidyl-POSS®

  • CAS Number 230316-12-8

  • Empirical Formula (Hill Notation) C38H68O13Si8

  • Molecular Weight 957.62

  •  PubChem Substance ID 24871392

Purchase

Properties

Related Categories Completely Condensed, Materials Science, Nanomaterials, Silsesquioxanes: POSS Nanohybrids
InChI Key   IDQZDTXGTHNXFY-UHFFFAOYSA-N
assay   95%
mp   >350 °C (lit.)

Description

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?