• USA Home
  • 477664 - PSS-(1-(Methyl propionate))-Heptacyclopentyl substituted

EMAIL THIS PAGE TO A FRIEND
477664 Sigma-Aldrich

PSS-(1-(Methyl propionate))-Heptacyclopentyl substituted

Synonym: Methyl 3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane-1-propionate, Methyl propionate-POSS®

Purchase

Properties

mp   >350 °C (lit.)
SMILES string   COC(=O)CC[Si]12O[Si]3(O[Si]4(O[Si](O1)(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)C6CCCC6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)C%11CCCC%11)C%12CCCC%12

Description

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?