EMAIL THIS PAGE TO A FRIEND
47816 Aldrich

Fmoc-D-Phe(2-Me)-OH

≥98.0% (HPLC)

Synonym: Fmoc-2-methyl-D-phenylalanine

  • Empirical Formula (Hill Notation) C25H23NO4

  • Molecular Weight 401.45

  •  MDL number MFCD00671404

  •  PubChem Substance ID 24871464

Purchase

Properties

assay   ≥98.0% (HPLC)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?