• USA Home
  • 478288 - 4-Chloro-2-nitrophenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
478288 Sigma-Aldrich

4-Chloro-2-nitrophenyl isocyanate

97%

Purchase

Properties

assay   97%
mp   65-68 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?