• USA Home
  • 478490 - Tolylene 2,5-diisocyanate

EMAIL THIS PAGE TO A FRIEND
478490 Sigma-Aldrich

Tolylene 2,5-diisocyanate

98%

Purchase

Properties

assay   98%
bp   138-139 °C/15 mmHg (lit.)
mp   35-39 °C (lit.)
SMILES string   Cc1cc(ccc1N=C=O)N=C=O
InChI   1S/C9H6N2O2/c1-7-4-8(10-5-12)2-3-9(7)11-6-13/h2-4H,1H3
InChI key   DKBHJZFJCDOGOY-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
WGK 3
Flash Point(F) 
235.4 °F - closed cup
Flash Point(C) 
113 °C - closed cup
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?