• USA Home
  • 479802 - Dichloro(dimethylglyoximato)(dimethylglyoxime)rhodium(III)

EMAIL THIS PAGE TO A FRIEND
479802 Sigma-Aldrich

Dichloro(dimethylglyoximato)(dimethylglyoxime)rhodium(III)

97%

  • CAS Number 39174-43-1

  • Empirical Formula (Hill Notation) C8H15Cl2N4O4Rh

  • Molecular Weight 405.04

  •  MDL number MFCD01321356

  •  PubChem Substance ID 24871651

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Rhodium Catalysts More...
InChI Key   IXIZJXPUAJCWJO-KMGPWXGDSA-K
assay   97%
mp   290 °C (dec.) (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?