• USA Home
  • 480614 - N,N-Bis(2-hydroxyethyl)ethylenediamine

EMAIL THIS PAGE TO A FRIEND
480614 Aldrich

N,N-Bis(2-hydroxyethyl)ethylenediamine

95%

Purchase

Properties

assay   95%
refractive index   n20/D 1.498 (lit.)
bp   110 °C/0.02 mmHg(lit.)
density   1.069 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN1 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?