• USA Home
  • 480711 - 6-O-(tert-Butyldiphenylsilyl)-D-galactal

EMAIL THIS PAGE TO A FRIEND
480711 Aldrich

6-O-(tert-Butyldiphenylsilyl)-D-galactal

97%

Purchase

Properties

InChI Key   BSEPYXNJJUUAEY-BQJUDKOJSA-N
assay   97%
optical activity   [α]20/D +4°, c = 1 in chloroform
contains   ~5 wt. % ethyl acetate to induce mobility of syrup

Description

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?