• USA Home
  • 480924 - Yttrium(III) oxalate hydrate

EMAIL THIS PAGE TO A FRIEND
480924 Sigma-Aldrich

Yttrium(III) oxalate hydrate

99.99%

  • CAS Number 252900-41-7

  • Linear Formula Y2(C2O4)3 · xH2O

  • Molecular Weight 441.87 (anhydrous basis)

  •  MDL number MFCD00150500

Purchase

Properties

assay   99.99%
mp   >400 °C (lit.)
density   2.51 g/mL at 25 °C (lit.)
SMILES string   O.O=C1O[Y](OC1=O)OC(=O)C(=O)O[Y]2OC(=O)C(=O)O2
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?