EMAIL THIS PAGE TO A FRIEND
481149 Aldrich

Silver(I) sulfite

99.99% trace metals basis

Purchase

Properties

assay   99.99% trace metals basis
form   solid
mp   100 °C (dec.)(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?