• USA Home
  • 481203 - Bis(isopropylcyclopentadienyl)zirconium(IV) dichloride

EMAIL THIS PAGE TO A FRIEND
481203 Aldrich

Bis(isopropylcyclopentadienyl)zirconium(IV) dichloride

98%

  • CAS Number 58628-40-3

  • Empirical Formula (Hill Notation) C16H22Cl2Zr

  • Molecular Weight 376.48

  •  MDL number MFCD00269855

  •  PubChem Substance ID 24871747

Purchase

Properties

assay   98%
mp   134-137 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?