EMAIL THIS PAGE TO A FRIEND
482080 Sigma-Aldrich

2,3-Dihydrofuran

≥99%

Synonym: 2,3-DHF

  • CAS Number 1191-99-7

  • Empirical Formula (Hill Notation) C4H6O

  • Molecular Weight 70.09

  •  Beilstein Registry Number 103168

  •  EC Number 214-747-7

  •  MDL number MFCD00003205

Purchase

Properties

vapor pressure   14.46 psi ( 55 °C)
  3.67 psi ( 20 °C)
InChI Key   JKTCBAGSMQIFNL-UHFFFAOYSA-N
assay   ≥99%
refractive index   n20/D 1.423(lit.)
bp   54-55 °C(lit.)
density   0.927 g/mL at 25 °C(lit.)

Description

Legal Information

Manufactured by Eastman Chemical Company. Distributed by Aldrich Chemical Company.

Safety & Documentation

Safety Information

RIDADR 
UN 1993 3/PG 2
WGK Germany 
3
Flash Point(F) 
-4 °F
Flash Point(C) 
-20 °C
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

200018 2,3-Dihydrofuran, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?