• USA Home
  • 482870 - (S)-(+)-1-(2-Naphthyl)-1,2-ethanediol

EMAIL THIS PAGE TO A FRIEND
482870 Sigma-Aldrich

(S)-(+)-1-(2-Naphthyl)-1,2-ethanediol

98%

  • CAS Number 43210-74-8

  • Linear Formula C10H7CH(OH)CH2OH

  • Molecular Weight 188.22

  •  PubChem Substance ID 24871852

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   QFNOPZNUHSDHKZ-GFCCVEGCSA-N
assay   98%
optical activity   [α]23/D +34°, c = 1 in ethanol
mp   131-135 °C (lit.)

Description

Packaging

500 mg in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?