• USA Home
  • 482935 - (R,R)-(−)-1-Phenylcyclohexane-cis-1,2-diol

EMAIL THIS PAGE TO A FRIEND
482935 Sigma-Aldrich

(R,R)-(−)-1-Phenylcyclohexane-cis-1,2-diol

99%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
assay   99%
optical activity   [α]23/D −6°, c = 1 in methanol
mp   121-123 °C (lit.)
SMILES string   O[C@@H]1CCCC[C@@]1(O)c2ccccc2
InChI   1S/C12H16O2/c13-11-8-4-5-9-12(11,14)10-6-2-1-3-7-10/h1-3,6-7,11,13-14H,4-5,8-9H2/t11-,12-/m1/s1
InChI key   QHNHEYDAIICUDL-VXGBXAGGSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?