• USA Home
  • 483427 - 1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane

EMAIL THIS PAGE TO A FRIEND
483427 Sigma-Aldrich

1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane

97%

Purchase

Properties

Related Categories Chemical Synthesis, Di-Alkoxy Silanes, Materials Science, Micro/NanoElectronics, Organometallic Reagents,
Quality Level   100
assay   97%
refractive index   n20/D 1.389 (lit.)
bp   161 °C (lit.)
density   0.883 g/mL at 25 °C (lit.)
SMILES string   CCO[Si](C)(C)O[Si](C)(C)OCC
InChI   1S/C8H22O3Si2/c1-7-9-12(3,4)11-13(5,6)10-8-2/h7-8H2,1-6H3
InChI key   NPOYZXWZANURMM-UHFFFAOYSA-N

Description

Packaging

25 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
109.4 °F - closed cup
Flash Point(C) 
43 °C - closed cup
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?