EMAIL THIS PAGE TO A FRIEND
483982 Sigma-Aldrich

Lithium bromide

beads, minimum melting composition, −10 mesh, ≥99.9%

Purchase

Properties

Related Categories 3: Li, Beaded Materials, Chemical Synthesis, Essential Chemicals, Inorganic Salts,
InChI Key   AMXOYNBUYSYVKV-UHFFFAOYSA-M
assay   ≥99.9%
form   beads
composition   LiBr, 68.5 wt. %
  LiCl, 22 wt. %
  LiF, 9.5 wt. %
particle size   −10 mesh
mp   550 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Complex Hydrides: A New Category of Solid-state Lithium Fast-ion Conductors

Research and development of solid-state lithium fast-ion conductors is crucial because they can be potentially used as solid electrolytes in all-solid-state batteries, which may solve the safety and ...
Motoaki Matsuo,1 Hiroyuki Oguchi,1,2 Hideki Maekawa,2 Hitoshi Takamura,2Shin-ichi Orimo,1*
Material Matters Volume 5 Article 4
Keywords: Calorimetry, Deposition, Diffraction, Diffusion, Melting, Nuclear magnetic resonance spectroscopy, Raman spectroscopy, Search, Spectra, Spectroscopy, Substitutions, Support, X-Ray diffraction, transformation

Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?