• USA Home
  • 486108 - Nitric-15N oxide-18O

EMAIL THIS PAGE TO A FRIEND
486108 Aldrich

Nitric-15N oxide-18O

98 atom % 15N, 95 atom % 18O

Purchase

Properties

Related Categories Chemical Synthesis, Gases, Gases and Gas Mixtures, Specialty Gases, Stable Isotopes,
vapor density   1.05 (vs air)
InChI Key   MWUXSHHQAYIFBG-RGIGPVFXSA-N
isotopic purity   98 atom % 15N
  95 atom % 18O
form   gas
bp   −151.7 °C(lit.)
mp   −163.6 °C(lit.)
mass shift   M+3

Description

Packaging

1 L in stainless steel cylinder

1 L quantity is packaged in a 450 mL stainless steel lecture bottle with stainless steel 1/4" male NPT valve.

This product cannot be packaged in different cylinder or with different valve.

Nominal gas pressures at 21ºC is 20 psig for 1 L. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.

This product available in U.S. only.

Recommended products

PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1660 8(5.1)(2.3)
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Custom Gas Packaging