• USA Home
  • 487252 - 2-Butyn-1-yl chloroformate

EMAIL THIS PAGE TO A FRIEND
487252 Sigma-Aldrich

2-Butyn-1-yl chloroformate

98%

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
assay   98%
refractive index   n20/D 1.452 (lit.)
bp   156 °C (lit.)
density   1.169 g/mL at 25 °C (lit.)
SMILES string   CC#CCOC(Cl)=O
InChI   1S/C5H5ClO2/c1-2-3-4-8-5(6)7/h4H2,1H3
InChI key   MFKKPMUEGDJJSL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3390 6.1/PG 1
WGK Germany 
3
Flash Point(F) 
140 °F
Flash Point(C) 
60 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?