EMAIL THIS PAGE TO A FRIEND
490377 Sigma-Aldrich

Nitrous oxide-15N2

98 atom % 15N

Synonym: Dinitrogen-15N2 monoxide

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, Chemical Synthesis, Gases, Gases and Gas Mixtures, N-O,
vapor density   1.53 (15 °C, vs air)
vapor pressure   51.7 mmHg ( 21 °C)
InChI Key   GQPLMRYTRLFLPF-ZDOIIHCHSA-N
isotopic purity   98 atom % 15N
bp   −88 °C(lit.)
mp   −91 °C (lit.)
mass shift   M+2

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

The use of a brass regulator is recommended.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1070 - class 2 - Nitrous oxide
WGK Germany 
1
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?