• USA Home
  • 490385 - [3aS-(3aα,4α,5α,7aα)]-3a,4,5,7a-Tetrahydro-2,2-dimethyl-1,3-benzodioxole-4,5-diol

EMAIL THIS PAGE TO A FRIEND
490385 Sigma-Aldrich

[3aS-(3aα,4α,5α,7aα)]-3a,4,5,7a-Tetrahydro-2,2-dimethyl-1,3-benzodioxole-4,5-diol

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
assay   98%
optical activity   [α]20/D −158°, c = 1 in chloroform
mp   68-71 °C (lit.)
SMILES string   [H][C@]12OC(C)(C)O[C@@]1([H])[C@H](O)[C@H](O)C=C2
InChI   1S/C9H14O4/c1-9(2)12-6-4-3-5(10)7(11)8(6)13-9/h3-8,10-11H,1-2H3/t5-,6-,7-,8-/m1/s1
InChI key   VCIXQRISGLSULF-WCTZXXKLSA-N

Description

Packaging

500 mg in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?