• USA Home
  • 492698 - Fmoc-Gly-OH-1-13C,15N

EMAIL THIS PAGE TO A FRIEND
492698 Sigma-Aldrich

Fmoc-Gly-OH-1-13C,15N

99 atom % 13C, 98 atom % 15N

Synonym: N-(9-Fluorenylmethoxycarbonyl)-glycine-1-13C,15N, Glycine-1-13C,15N, N-Fmoc

Purchase

Properties

Related Categories Alphabetical Listings, Amino Acids and Derivatives, Biomolecular NMR, E-F, Protected Amino Acids,
InChI Key   NDKDFTQNXLHCGO-ORUFPBKPSA-N
isotopic purity   99 atom % 13C
  98 atom % 15N
mp   174-175 °C (lit.)
mass shift   M+2
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?