• USA Home
  • 492760 - 2-Acetyl-7-methoxybenzofuran

EMAIL THIS PAGE TO A FRIEND
492760 Sigma-Aldrich

2-Acetyl-7-methoxybenzofuran

98%

Purchase

Properties

Related Categories Benzofurans, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
Quality Level   100
assay   98%
mp   94-96 °C (lit.)
SMILES string   COc1cccc2cc(oc12)C(C)=O
InChI   1S/C11H10O3/c1-7(12)10-6-8-4-3-5-9(13-2)11(8)14-10/h3-6H,1-2H3
InChI key   MUUMFANQVPWPFY-UHFFFAOYSA-N

Description

General description

2-Acetyl-7-methoxybenzofuran is a substituted benzofuran. It has been extracted as one of the metabolites from Sporothrix sp., a mangrove endophytic fungus.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?