• USA Home
  • 493600 - Bis(pentamethylcyclopentadienyl)nickel(II)

EMAIL THIS PAGE TO A FRIEND
493600 Sigma-Aldrich

Bis(pentamethylcyclopentadienyl)nickel(II)

Synonym: Decamethylnickelocene

Purchase

Properties

mp   261-266 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?