• USA Home
  • 493686 - Bis(tert-butylcyclopentadienyl)hafnium(IV) dichloride

EMAIL THIS PAGE TO A FRIEND
493686 Sigma-Aldrich

Bis(tert-butylcyclopentadienyl)hafnium(IV) dichloride

95%

  • CAS Number 33010-55-8

  • Empirical Formula (Hill Notation) C18H26Cl2Hf

  • Molecular Weight 491.80

  •  MDL number MFCD01073803

  •  PubChem Substance ID 24872852

Purchase

Properties

assay   95%
reaction suitability   reagent type: catalyst
core: hafnium
mp   195 °C (lit.)
SMILES string   Cl[Hf]Cl.CC(C)(C)[C]1[CH][CH][CH][CH]1.CC(C)(C)[C]2[CH][CH][CH][CH]2
InChI   1S/2C9H13.2ClH.Hf/c2*1-9(2,3)8-6-4-5-7-8;;;/h2*4-7H,1-3H3;2*1H;/q;;;;+2/p-2
InChI key   AJFWNXFQMFKIOR-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?