• USA Home
  • 493708 - Bis(tert-butylcyclopentadienyl)dimethylhafnium(IV)

EMAIL THIS PAGE TO A FRIEND
493708 Sigma-Aldrich

Bis(tert-butylcyclopentadienyl)dimethylhafnium(IV)

95%

Purchase

Properties

assay   95%
form   solid
reaction suitability   core: hafnium
mp   73-76 °C (lit.)
SMILES string   C[Hf]C.CC(C)(C)[C]1[CH][CH][CH][CH]1.CC(C)(C)[C]2[CH][CH][CH][CH]2
InChI   1S/2C9H13.2CH3.Hf/c2*1-9(2,3)8-6-4-5-7-8;;;/h2*4-7H,1-3H3;2*1H3;
InChI key   SOSWMGMRDGAPIG-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1325 - class 4.1 - PG 3 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?