• USA Home
  • 493740 - (S)-(−)-N-tert-Butyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxamide

EMAIL THIS PAGE TO A FRIEND
493740 Sigma-Aldrich

(S)-(−)-N-tert-Butyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxamide

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Isoquinolines More...
assay   98%
optical activity   [α]22/D −104°, c = 0.9 in methylene chloride
mp   98-99 °C (lit.)
SMILES string   CC(C)(C)NC(=O)[C@@H]1Cc2ccccc2CN1
InChI   1S/C14H20N2O/c1-14(2,3)16-13(17)12-8-10-6-4-5-7-11(10)9-15-12/h4-7,12,15H,8-9H2,1-3H3,(H,16,17)/t12-/m0/s1
InChI key   DMJXRYSGXCLCFP-LBPRGKRZSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?