• USA Home
  • 493961 - 4-Chlorophenyl benzoate

EMAIL THIS PAGE TO A FRIEND
493961 Sigma-Aldrich

4-Chlorophenyl benzoate

99%

Synonym: p -Chlorophenyl benzoate

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   99%
mp   87-89 °C (lit.)
SMILES string   Clc1ccc(OC(=O)c2ccccc2)cc1
InChI   1S/C13H9ClO2/c14-11-6-8-12(9-7-11)16-13(15)10-4-2-1-3-5-10/h1-9H
InChI key   JKSIXXOEIXUYFW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?