• USA Home
  • 494399 - 1,1,2,2,3,4-Hexafluoro-3,4-bis(trifluoromethyl)cyclobutane, mixture of cis and trans

EMAIL THIS PAGE TO A FRIEND
494399 Sigma-Aldrich

1,1,2,2,3,4-Hexafluoro-3,4-bis(trifluoromethyl)cyclobutane, mixture of cis and trans

95%

Purchase

Properties

assay   95%
impurities   ≤5% 2,4-isomer
refractive index   n20/D 1.3 (lit.)
bp   45 °C (lit.)
mp   −32 °C (lit.)
density   1.67 g/mL at 25 °C (lit.)
SMILES string   FC(F)(F)C1(F)C(F)(F)C(F)(F)C1(F)C(F)(F)F
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?