• USA Home
  • 494933 - Poly(ethylene adipate)

EMAIL THIS PAGE TO A FRIEND
494933 Sigma-Aldrich

Poly(ethylene adipate)

average Mn ~1,000

Synonym: Ethylene glycol adipate, Poly(ethylene adipate), dihydroxy terminated

Purchase

Properties

mol wt   average Mn ~1,000
viscosity   300 cP(60 °C)(lit.)
density   1.175 g/mL at 25 °C
InChI   1S/C6H10O4.C2H5O2/c7-5(8)3-1-2-4-6(9)10;3-1-2-4/h1-4H2,(H,7,8)(H,9,10);3H,1-2H2
InChI key   CODQTHDBFINLJQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?